Assay Details
| Gene Symbol: cry5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131788.1 | NP_571863.1 | 6 - 7 | 991 | 99 | |
| XM_005155405.3 | XP_005155462.1 | 7 - 8 | 99 | |||
| GenBank mRNA | AB042254.1 | - | 6 - 7 | 991 | 99 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

cry5
Genomic Map