Assay Details
| Gene Symbol: lmna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152971.1 | NP_694503.1 | 10 - 11 | 1727 | 67 | |
| XM_005158167.4 | XP_005158224.1 | 11 - 12 | 2116 | 67 | ||
| XM_005158168.4 | XP_005158225.1 | 11 - 12 | 1845 | 67 | ||
| GenBank mRNA | AF397016.1 | - | 10 - 11 | 1727 | 67 | |
| BC163799.1 | - | 10 - 11 | 1750 | 67 | ||
| BC163807.1 | - | 10 - 11 | 1750 | 67 | ||
More Information
Set Membership: |
|

lmna
Genomic Map