Assay Details
| Gene Symbol: npffl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183346.3 | NP_899189.2 | 1 - 2 | 174 | 105 | |
| GenBank mRNA | AB193140.1 | - | 1 - 2 | 174 | 105 | |
| BC094983.1 | - | 1 - 2 | 192 | 105 | ||
More Information
Set Membership: |
|

npffl
Genomic Map