Assay Details
| Gene Symbol: fen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328507.1 | NP_001315436.1 | 5 - 6 | 485 | 106 | |
| XM_017353804.1 | XP_017209293.1 | 5 - 6 | 503 | 106 | ||
| GenBank mRNA | AY391423.1 | - | 5 - 6 | 414 | 106 | |
| AY648844.1 | - | 5 - 6 | 427 | 106 | ||
| BC049413.1 | - | 5 - 6 | 424 | 106 | ||
| BC071488.1 | - | 5 - 6 | 446 | 106 | ||
| CT612850.2 | - | 5 - 6 | 464 | 106 | ||
| EH439441.1 | - | 5 - 6 | 493 | 106 | ||
More Information
Set Membership: |
|

fen1
Genomic Map