Assay Details
| Gene Symbol: tmed2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199548.2 | NP_955842.1 | 1 - 2 | 310 | 106 | |
| XM_021479108.1 | XP_021334783.1 | 1 - 2 | 316 | 106 | ||
| GenBank mRNA | BC050165.1 | - | 1 - 2 | 279 | 106 | |
| BC065441.1 | - | 1 - 2 | 251 | 106 | ||
| EB783930.1 | - | 1 - 2 | 314 | 106 | ||
More Information
Set Membership: |
|

tmed2
Genomic Map