Assay Details
Gene Symbol: abi3bpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199859.1 | NP_956153.1 | 15 - 16 | 1861 | 113 | |
XM_005161776.4 | XP_005161833.1 | 16 - 17 | 1939 | 113 | ||
XM_005161777.4 | XP_005161834.1 | 15 - 16 | 1738 | 113 | ||
XM_009296432.3 | XP_009294707.1 | 14 - 15 | 1705 | 113 | ||
XM_009296433.3 | XP_009294708.1 | 14 - 15 | 1663 | 113 | ||
XM_009296434.3 | XP_009294709.1 | 13 - 14 | 1591 | 113 | ||
XM_009296436.3 | XP_009294711.1 | 12 - 13 | 1429 | 113 | ||
XM_009296437.3 | XP_009294712.1 | 12 - 13 | 1423 | 113 | ||
XM_009296438.3 | XP_009294713.1 | 11 - 12 | 1357 | 113 | ||
XM_021469291.1 | XP_021324966.1 | 15 - 16 | 1864 | 113 | ||
XM_021469292.1 | XP_021324967.1 | 13 - 14 | 1495 | 113 | ||
GenBank mRNA | BC045333.1 | - | 15 - 16 | 1861 | 113 | |
BC165666.1 | - | 15 - 16 | 1778 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
