Assay Details
Gene Symbol: abi3bpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199859.1 | NP_956153.1 | 16 - 17 | 1967 | 74 | |
XM_005161776.4 | XP_005161833.1 | 17 - 18 | 2045 | 74 | ||
XM_005161777.4 | XP_005161834.1 | 16 - 17 | 1844 | 74 | ||
XM_009296432.3 | XP_009294707.1 | 15 - 16 | 1811 | 74 | ||
XM_009296433.3 | XP_009294708.1 | 15 - 16 | 1769 | 74 | ||
XM_009296434.3 | XP_009294709.1 | 14 - 15 | 1697 | 74 | ||
XM_009296436.3 | XP_009294711.1 | 13 - 14 | 1535 | 74 | ||
XM_009296437.3 | XP_009294712.1 | 13 - 14 | 1529 | 74 | ||
XM_009296438.3 | XP_009294713.1 | 12 - 13 | 1463 | 74 | ||
XM_021469291.1 | XP_021324966.1 | 16 - 17 | 1970 | 74 | ||
XM_021469292.1 | XP_021324967.1 | 14 - 15 | 1601 | 74 | ||
GenBank mRNA | BC045333.1 | - | 16 - 17 | 1967 | 74 | |
BC165666.1 | - | 16 - 17 | 1884 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
