Assay Details
Gene Symbol: abi3bpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199859.1 | NP_956153.1 | 17 - 18 | 2046 | 83 | |
XM_005161776.4 | XP_005161833.1 | 18 - 19 | 2124 | 83 | ||
XM_005161777.4 | XP_005161834.1 | 17 - 18 | 1923 | 83 | ||
XM_009296432.3 | XP_009294707.1 | 16 - 17 | 1890 | 83 | ||
XM_009296433.3 | XP_009294708.1 | 16 - 17 | 1848 | 83 | ||
XM_009296434.3 | XP_009294709.1 | 15 - 16 | 1776 | 83 | ||
XM_009296436.3 | XP_009294711.1 | 14 - 15 | 1614 | 83 | ||
XM_009296437.3 | XP_009294712.1 | 14 - 15 | 1608 | 83 | ||
XM_009296438.3 | XP_009294713.1 | 13 - 14 | 1542 | 83 | ||
XM_021469291.1 | XP_021324966.1 | 17 - 18 | 2049 | 83 | ||
XM_021469292.1 | XP_021324967.1 | 15 - 16 | 1680 | 83 | ||
GenBank mRNA | BC045333.1 | - | 17 - 18 | 2046 | 83 | |
BC165666.1 | - | 17 - 18 | 1963 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
