Assay Details
Gene Symbol: abi3bpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199859.1 | NP_956153.1 | 1 - 2 | 211 | 62 | |
XM_005161776.4 | XP_005161833.1 | 1 - 2 | 223 | 62 | ||
XM_005161777.4 | XP_005161834.1 | 1 - 2 | 223 | 62 | ||
XM_009296432.3 | XP_009294707.1 | 1 - 2 | 223 | 62 | ||
XM_009296433.3 | XP_009294708.1 | 1 - 2 | 223 | 62 | ||
XM_009296434.3 | XP_009294709.1 | 1 - 2 | 223 | 62 | ||
XM_009296436.3 | XP_009294711.1 | 1 - 2 | 223 | 62 | ||
XM_009296437.3 | XP_009294712.1 | 1 - 2 | 223 | 62 | ||
XM_009296438.3 | XP_009294713.1 | 1 - 2 | 223 | 62 | ||
XM_021469291.1 | XP_021324966.1 | 1 - 2 | 223 | 62 | ||
XM_021469292.1 | XP_021324967.1 | 1 - 2 | 223 | 62 | ||
GenBank mRNA | AW175067.1 | - | 1 - 2 | 205 | 62 | |
BC045333.1 | - | 1 - 2 | 211 | 62 | ||
BC165666.1 | - | 1 - 2 | 128 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
