Assay Details
Gene Symbol: abi3bpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199859.1 | NP_956153.1 | 6 - 7 | 825 | 112 | |
XM_005161776.4 | XP_005161833.1 | 6 - 7 | 837 | 112 | ||
XM_005161777.4 | XP_005161834.1 | 6 - 7 | 837 | 112 | ||
XM_009296432.3 | XP_009294707.1 | 6 - 7 | 837 | 112 | ||
XM_009296433.3 | XP_009294708.1 | 6 - 7 | 837 | 112 | ||
XM_009296434.3 | XP_009294709.1 | 6 - 7 | 837 | 112 | ||
XM_009296436.3 | XP_009294711.1 | 6 - 7 | 837 | 112 | ||
XM_009296437.3 | XP_009294712.1 | 6 - 7 | 837 | 112 | ||
XM_009296438.3 | XP_009294713.1 | 6 - 7 | 837 | 112 | ||
XM_021469291.1 | XP_021324966.1 | 6 - 7 | 837 | 112 | ||
XM_021469292.1 | XP_021324967.1 | 6 - 7 | 837 | 112 | ||
GenBank mRNA | BC045333.1 | - | 6 - 7 | 825 | 112 | |
BC165666.1 | - | 6 - 7 | 742 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
