Assay Details
| Gene Symbol: vamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200005.1 | NP_956299.1 | 4 - 5 | 543 | 90 | |
| GenBank mRNA | BC059626.1 | - | 4 - 5 | 543 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

vamp2| Gene Symbol: vamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200005.1 | NP_956299.1 | 4 - 5 | 543 | 90 | |
| GenBank mRNA | BC059626.1 | - | 4 - 5 | 543 | 90 | |
Set Membership: |
|