Assay Details
Gene Symbol: ubl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200360.1 | NP_956654.1 | 8 - 9 | 775 | 107 | |
XM_005173685.3 | XP_005173742.1 | 8 - 9 | 827 | 107 | ||
XM_005173686.4 | XP_005173743.1 | 8 - 9 | 827 | 107 | ||
XM_009293893.3 | XP_009292168.1 | 8 - 9 | 839 | 107 | ||
GenBank mRNA | BC053194.1 | - | 8 - 9 | 775 | 107 | |
BC164195.1 | - | 7 - 8 | 733 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
