Assay Details
| Gene Symbol: atp5pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200702.1 | NP_956996.1 | 3 - 4 | 256 | 57 | |
| XM_005163774.4 | XP_005163831.1 | 3 - 4 | 285 | 57 | ||
| GenBank mRNA | BC059446.1 | - | 3 - 4 | 256 | 57 | |
| BC150460.1 | - | 3 - 4 | 267 | 57 | ||
More Information
Set Membership: |
|

atp5pd
Genomic Map