Assay Details
Gene Symbol: skp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200743.1 | NP_957037.1 | 2 - 3 | 142 | 63 | |
GenBank mRNA | AY648843.1 | - | 1 - 2 | 122 | 63 | |
BC059536.1 | - | 2 - 3 | 142 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
