Assay Details
Gene Symbol: fkbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200812.2 | NP_957106.1 | 2 - 3 | 169 | 69 | |
GenBank mRNA | BC059689.1 | - | 2 - 3 | 159 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
