Assay Details
| Gene Symbol: pdk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200996.1 | NP_957290.1 | 5 - 6 | 717 | 84 | |
| XM_021468022.1 | XP_021323697.1 | 5 - 6 | 84 | |||
| GenBank mRNA | BC045993.1 | - | 5 - 6 | 717 | 84 | |
More Information
Set Membership: |
|

pdk2b
Genomic Map