Assay Details
| Gene Symbol: rpl18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201060.1 | NP_957354.1 | 3 - 4 | 365 | 111 | |
| GenBank mRNA | BC049045.1 | - | 3 - 4 | 365 | 111 | |
More Information
Set Membership: |
|

rpl18a
Genomic Map