Assay Details
Gene Symbol: atp6v1aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201135.2 | NP_957429.1 | 9 - 10 | 1174 | 79 | |
GenBank mRNA | BC055130.1 | - | 9 - 10 | 1175 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
