Assay Details
Gene Symbol: asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201163.3 | NP_957457.3 | 4 - 5 | 739 | 79 | |
GenBank mRNA | BC052127.1 | - | 4 - 5 | 749 | 79 | |
BC067140.1 | - | 4 - 5 | 739 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
