Assay Details
| Gene Symbol: rab5aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201485.1 | NP_958893.1 | 3 - 4 | 507 | 65 | |
| GenBank mRNA | BC047803.1 | - | 3 - 4 | 507 | 65 | |
| BC063966.1 | - | 3 - 4 | 502 | 65 | ||
| BC165915.1 | - | 2 - 3 | 347 | 65 | ||
More Information
Set Membership: |
|

rab5aa
Genomic Map