Assay Details
| Gene Symbol: ilf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212599.1 | NP_997764.1 | 13 - 14 | 1639 | 86 | |
| XM_005163739.4 | XP_005163796.1 | 13 - 14 | 1711 | 86 | ||
| XM_005163740.4 | XP_005163797.1 | 13 - 14 | 1711 | 86 | ||
| XM_005163741.3 | XP_005163798.1 | 13 - 14 | 1708 | 86 | ||
| XM_005163742.4 | XP_005163799.1 | 13 - 14 | 1708 | 86 | ||
| XM_009299225.3 | XP_009297500.1 | 12 - 13 | 1619 | 86 | ||
| GenBank mRNA | BC067172.1 | - | 13 - 14 | 1639 | 86 | |
More Information
Set Membership: |
|

ilf3b
Genomic Map