Assay Details
| Gene Symbol: ilf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212599.1 | NP_997764.1 | 9 - 10 | 1056 | 56 | |
| XM_005163739.4 | XP_005163796.1 | 9 - 10 | 1128 | 56 | ||
| XM_005163740.4 | XP_005163797.1 | 9 - 10 | 1128 | 56 | ||
| XM_005163741.3 | XP_005163798.1 | 9 - 10 | 1125 | 56 | ||
| XM_005163742.4 | XP_005163799.1 | 9 - 10 | 1125 | 56 | ||
| XM_009299225.3 | XP_009297500.1 | 8 - 9 | 1036 | 56 | ||
| GenBank mRNA | BC047175.1 | - | 9 - 10 | 1085 | 56 | |
| BC067172.1 | - | 9 - 10 | 1056 | 56 | ||
More Information
Set Membership: |
|

ilf3b
Genomic Map