Assay Details
| Gene Symbol: spint1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213152.1 | NP_998317.1 | 3 - 4 | 720 | 133 | |
| XM_021467136.1 | XP_021322811.1 | 4 - 5 | 808 | 133 | ||
| XM_021467137.1 | XP_021322812.1 | 4 - 5 | 809 | 133 | ||
| GenBank mRNA | BC053239.1 | - | 3 - 4 | 720 | 133 | |
| BC152670.1 | - | 3 - 4 | 748 | 133 | ||
| BC155204.1 | - | 3 - 4 | 669 | 133 | ||
| BC165390.1 | - | 2 - 3 | 597 | 133 | ||
More Information
Set Membership: |
|

spint1a
Genomic Map