Assay Details
| Gene Symbol: pitpnbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213443.1 | NP_998608.1 | 3 - 4 | 338 | 105 | |
| GenBank mRNA | BC046882.1 | - | 3 - 4 | 338 | 105 | |
| BC164619.1 | - | 3 - 4 | 237 | 105 | ||
More Information
Set Membership: |
|

pitpnbl
Genomic Map