Assay Details
| Gene Symbol: pitpnbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213443.1 | NP_998608.1 | 6 - 7 | 506 | 122 | |
| GenBank mRNA | BC046882.1 | - | 6 - 7 | 506 | 122 | |
| BC164619.1 | - | 6 - 7 | 405 | 122 | ||
More Information
Set Membership: |
|

pitpnbl
Genomic Map