Assay Details
| Gene Symbol: sh3bp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213458.1 | NP_998623.1 | 4 - 5 | 590 | 99 | |
| GenBank mRNA | BC044426.1 | - | 4 - 5 | 590 | 99 | |
| BC165200.1 | - | 4 - 5 | 466 | 99 | ||
More Information
Set Membership: |
|

sh3bp5b
Genomic Map