Assay Details
Gene Symbol: hspa4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214716.1 | NP_999881.1 | 17 - 18 | 2345 | 91 | |
GenBank mRNA | BC051152.1 | - | 17 - 18 | 2345 | 91 | |
BC165369.1 | - | 17 - 18 | 2195 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
