Assay Details
Gene Symbol: psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002124.2 | NP_001002124.1 | 7 - 8 | 675 | 67 | |
GenBank mRNA | BC071453.1 | - | 7 - 8 | 677 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
