Assay Details
| Gene Symbol: rap1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002152.1 | NP_001002152.1 | 2 - 3 | 168 | 148 | |
| XM_005167023.3 | XP_005167080.1 | 2 - 3 | 228 | 148 | ||
| GenBank mRNA | BC049419.1 | - | 2 - 3 | 162 | 148 | |
| BC071360.1 | - | 2 - 3 | 168 | 148 | ||
More Information
Set Membership: |
|

rap1aa
Genomic Map