Assay Details
| Gene Symbol: pdha1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002399.1 | NP_001002399.1 | 7 - 8 | 825 | 87 | |
| XM_005162686.4 | XP_005162743.1 | 8 - 9 | 826 | 87 | ||
| GenBank mRNA | BC076185.1 | - | 7 - 8 | 825 | 87 | |
More Information
Set Membership: |
|

pdha1b
Genomic Map