Assay Details
| Gene Symbol: dgat1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002458.1 | NP_001002458.1 | 10 - 11 | 1163 | 124 | |
| XM_021466677.1 | XP_021322352.1 | 9 - 10 | 793 | 124 | ||
| GenBank mRNA | BC076012.1 | - | 10 - 11 | 1163 | 124 | |
More Information
Set Membership: |
|

dgat1b
Genomic Map