Assay Details
Gene Symbol: atp2b3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002472.3 | NP_001002472.2 | 11 - 12 | 2810 | 88 | |
NM_001365433.1 | NP_001352362.1 | 12 - 13 | 2876 | 88 | ||
XM_005166818.4 | XP_005166875.1 | 12 - 13 | 2795 | 88 | ||
XM_005166819.4 | XP_005166876.1 | 11 - 12 | 2728 | 88 | ||
XM_005166820.4 | XP_005166877.1 | 12 - 13 | 2795 | 88 | ||
XM_005166822.4 | XP_005166879.1 | 12 - 13 | 2797 | 88 | ||
XM_021478239.1 | XP_021333914.1 | 11 - 12 | 2728 | 88 | ||
XM_021478240.1 | XP_021333915.1 | 11 - 12 | 2730 | 88 | ||
GenBank mRNA | BC076342.1 | - | 10 - 11 | 2163 | 88 | |
EU559283.1 | - | 10 - 11 | 2159 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
