Assay Details
| Gene Symbol: n6amt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002502.1 | NP_001002502.1 | 6 - 7 | 575 | 67 | |
| GenBank mRNA | BC076297.1 | - | 6 - 7 | 575 | 67 | |
More Information
Set Membership: |
|

n6amt1
Genomic Map