Assay Details
Gene Symbol: itm2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002549.1 | NP_001002549.1 | 1 - 2 | 207 | 83 | |
GenBank mRNA | BC076257.1 | - | 1 - 2 | 207 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
