Assay Details
| Gene Symbol: hpdb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003742.1 | NP_001003742.1 | 11 - 12 | 873 | 66 | |
| GenBank mRNA | BC077167.1 | - | 11 - 12 | 873 | 66 | |
| BC165880.1 | - | 10 - 11 | 864 | 66 | ||
More Information
Set Membership: |
|

hpdb
Genomic Map