Assay Details
| Gene Symbol: hpdb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003742.1 | NP_001003742.1 | 13 - 14 | 1116 | 108 | |
| GenBank mRNA | BC077167.1 | - | 13 - 14 | 1116 | 108 | |
| BC165880.1 | - | 12 - 13 | 1107 | 108 | ||
More Information
Set Membership: |
|

hpdb
Genomic Map