Assay Details
Gene Symbol: cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003878.2 | NP_001003878.2 | 3 - 4 | 494 | 133 | |
GenBank mRNA | AY648825.1 | - | 3 - 4 | 506 | 133 | |
BC095551.1 | - | 3 - 4 | 494 | 133 | ||
EH592323.1 | - | 3 - 4 | 770 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
