Assay Details
| Gene Symbol: stk25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320056.1 | NP_001306985.1 | 10 - 11 | 1500 | 77 | |
| GenBank mRNA | BC083536.1 | - | 10 - 11 | 1342 | 77 | |
| EH524747.1 | - | 1 - 2 | 423 | 77 | ||
More Information
Set Membership: |
|

stk25a
Genomic Map