Assay Details
| Gene Symbol: bcl7bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006018.1 | NP_001006018.1 | 1 - 2 | 142 | 86 | |
| GenBank mRNA | BC083405.1 | - | 1 - 2 | 142 | 86 | |
| BC165913.1 | - | 1 - 2 | 122 | 86 | ||
More Information
Set Membership: |
|

bcl7bb
Genomic Map