Assay Details
| Gene Symbol: bcl7bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006018.1 | NP_001006018.1 | 5 - 6 | 589 | 150 | |
| GenBank mRNA | BC083405.1 | - | 5 - 6 | 589 | 150 | |
More Information
Set Membership: |
|

bcl7bb
Genomic Map