Assay Details
Gene Symbol: hrasa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017623.1 | NP_001017623.1 | 3 - 4 | 426 | 84 | |
GenBank mRNA | BC092826.1 | - | 3 - 4 | 426 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
