Assay Details
| Gene Symbol: hrasa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017623.1 | NP_001017623.1 | 5 - 6 | 710 | 71 | |
| GenBank mRNA | BC092826.1 | - | 5 - 6 | 710 | 71 | |
More Information
Set Membership: |
|

hrasa
Genomic Map