Assay Details
| Gene Symbol: arfip2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017649.1 | NP_001017649.1 | 8 - 9 | 1134 | 90 | |
| XM_005155373.4 | XP_005155430.1 | 9 - 10 | 1305 | 90 | ||
| XM_005155375.4 | XP_005155432.1 | 9 - 10 | 1280 | 90 | ||
| XM_009305311.3 | XP_009303586.1 | 9 - 10 | 1271 | 90 | ||
| XM_009305312.3 | XP_009303587.1 | 8 - 9 | 1247 | 90 | ||
| XM_009305313.3 | XP_009303588.1 | 8 - 9 | 1214 | 90 | ||
| XM_009305314.3 | XP_009303589.1 | 8 - 9 | 1207 | 90 | ||
| XM_021479173.1 | XP_021334848.1 | 7 - 8 | 1149 | 90 | ||
| GenBank mRNA | BC093181.1 | - | 8 - 9 | 1134 | 90 | |
| BC164396.1 | - | 7 - 8 | 948 | 90 | ||
More Information
Set Membership: |
|

arfip2b
Genomic Map