Assay Details
| Gene Symbol: syt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017652.1 | NP_001017652.1 | 7 - 8 | 1265 | 71 | |
| XM_017356856.2 | XP_017212345.2 | 5 - 6 | 1345 | 71 | ||
| GenBank mRNA | BC093178.1 | - | 7 - 8 | 1265 | 71 | |
More Information
Set Membership: |
|

syt13
Genomic Map