Assay Details
Gene Symbol: sema6dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017682.2 | NP_001017682.2 | 4 - 5 | 386 | 89 | |
XM_005173648.4 | XP_005173705.1 | 4 - 5 | 504 | 89 | ||
XM_005173649.4 | XP_005173706.1 | 4 - 5 | 504 | 89 | ||
XM_005173650.4 | XP_005173707.1 | 4 - 5 | 504 | 89 | ||
XM_009293805.3 | XP_009292080.1 | 4 - 5 | 895 | 89 | ||
XM_009293807.3 | XP_009292082.1 | 4 - 5 | 504 | 89 | ||
GenBank mRNA | AB212073.1 | - | 4 - 5 | 386 | 89 | |
AB212074.1 | - | 4 - 5 | 386 | 89 | ||
BC093159.1 | - | 4 - 5 | 553 | 89 | ||
CU458860.1 | - | 3 - 4 | 286 | 89 | ||
FJ209036.1 | - | 3 - 4 | 327 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
