Assay Details
Gene Symbol: sema6dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017682.2 | NP_001017682.2 | 5 - 6 | 447 | 146 | |
XM_005173648.4 | XP_005173705.1 | 5 - 6 | 565 | 146 | ||
XM_005173649.4 | XP_005173706.1 | 5 - 6 | 565 | 146 | ||
XM_005173650.4 | XP_005173707.1 | 5 - 6 | 565 | 146 | ||
XM_009293805.3 | XP_009292080.1 | 5 - 6 | 956 | 146 | ||
XM_009293807.3 | XP_009292082.1 | 5 - 6 | 565 | 146 | ||
GenBank mRNA | AB212073.1 | - | 5 - 6 | 447 | 146 | |
AB212074.1 | - | 5 - 6 | 447 | 146 | ||
BC093159.1 | - | 5 - 6 | 614 | 146 | ||
CU458858.1 | - | 4 - 5 | 353 | 146 | ||
CU458860.1 | - | 4 - 5 | 347 | 146 | ||
FJ209036.1 | - | 4 - 5 | 388 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
