Assay Details
Gene Symbol: sema6dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017682.2 | NP_001017682.2 | 7 - 8 | 643 | 63 | |
XM_005173648.4 | XP_005173705.1 | 7 - 8 | 761 | 63 | ||
XM_005173649.4 | XP_005173706.1 | 7 - 8 | 761 | 63 | ||
XM_005173650.4 | XP_005173707.1 | 7 - 8 | 761 | 63 | ||
XM_009293805.3 | XP_009292080.1 | 7 - 8 | 1152 | 63 | ||
XM_009293807.3 | XP_009292082.1 | 7 - 8 | 761 | 63 | ||
GenBank mRNA | AB212073.1 | - | 7 - 8 | 643 | 63 | |
AB212074.1 | - | 7 - 8 | 643 | 63 | ||
CU458858.1 | - | 6 - 7 | 549 | 63 | ||
CU458860.1 | - | 6 - 7 | 543 | 63 | ||
FJ209036.1 | - | 6 - 7 | 584 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
