Assay Details
| Gene Symbol: gpd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017709.1 | NP_001017709.1 | 7 - 8 | 1015 | 93 | |
| GenBank mRNA | BC093259.1 | - | 7 - 8 | 1015 | 93 | |
More Information
Set Membership: |
|

gpd1a
Genomic Map