Assay Details
| Gene Symbol: rgs5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017821.2 | NP_001017821.1 | 2 - 3 | 261 | 74 | |
| GenBank mRNA | BC092795.1 | - | 2 - 3 | 262 | 74 | |
| BC165301.1 | - | 2 - 3 | 215 | 74 | ||
More Information
Set Membership: |
|

rgs5b
Genomic Map