Assay Details
| Gene Symbol: nudt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017843.1 | NP_001017843.1 | 3 - 4 | 423 | 63 | |
| XM_005172467.3 | XP_005172524.1 | 3 - 4 | 463 | 63 | ||
| GenBank mRNA | BC092771.1 | - | 3 - 4 | 423 | 63 | |
More Information
Set Membership: |
|

nudt18
Genomic Map